missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Dimethylamino-Benzaldehyde, ACS, MP Biomedicals™
Click to view available options
Quantity:
50 g
100 g
250 g
Specifications
Quantity | 50 g |
Formula Weight | 149.2 |
Grade | ACS Reagent |
Physical Form | Solid |
Chemical Name or Material | p-Dimethylamino-Benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'