Läs mer
p-Hydroxy-(D)-phenylglycyl Amoxicillin (Technical Grade), TRC
Produktkod. 30137950
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137950 50 mg
30009067 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95085550MG
 Begränsad artikel
Beräknad leverans: 04-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 188112-75-6
Synonym Amoxicillin Trihydrate Imp. G (EP)∣D-(4-Hydroxyphenyl)-glycylamoxicillin∣(2R)-2-(4-hydroxyphenyl)glycyl-N-[(2S,5R,6R)-2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl]-2-(4-hydroxyphenyl)-Glycinamide
Specifikationer
Formatera Neat
Molekylvikt (g/mol) 514.55