Läs mer
p-Hydroxymandelamide, TRC
Produktkod. 30407495
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30407495 250 mg
1 options
Produktkod. 30407495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH944365250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-Hydroxymandelamide
CAS 74908-87-5
Molekylformel C8H9NO3
Kvantitet 250 mg
Synonym α,4-Dihydroxybenzeneacetamide, (±)-α,4-Dihydroxybenzeneacetamide
InChI formel InChI=1S/C8H9NO3/c9-8(12)7(11)5-1-3-6(10)4-2-5/h1-4,7,10-11H,(H2,9,12)
LEDER O=C(N)C(O)C1=CC=C(O)C=C1
Molekylvikt (g/mol) 167.16
Formel vikt 167.06