Läs mer
p-Methoxybenzyl Formate, TRC
Produktkod. 30465608
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30465608 10 mg
1 options
Produktkod. 30465608 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM57388010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-Methoxybenzyl Formate
CAS 122-91-8
Molekylformel C9H10O3
Kvantitet 10 mg
Synonym (4-Anisyl)methyl Formate, 4-Methoxybenzyl Formate, Anisyl Formate, NSC 5949
InChI formel InChI=1S/C9H10O3/c1-11-9-4-2-8(3-5-9)6-12-7-10/h2-5,7H,6H2,1H3
LEDER O=COCC1=CC=C(OC)C=C1
Molekylvikt (g/mol) 166.17