Läs mer
p-MPPF Hydrochloride, TRC
Produktkod. 30465003
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30465003 5 mg
1 options
Produktkod. 30465003 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM7458335MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-MPPF Hydrochloride
CAS 223699-41-0
Molekylformel C25H27FN4O2 • x(HCl)
Kvantitet 5 mg
Synonym 4-Fluoro-N-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinyl-benzamide Hydrochloride
InChI formel InChI=1S/C25H27FN4O2.2ClH/c1-32-23-7-3-2-6-22(23)29-17-14-28(15-18-29)16-19-30(24-8-4-5-13-27-24)25(31)20-9-11-21(26)12-10-20;;/h2-13H,14-19H2,1H3;2*1H
LEDER FC1=CC=C(C(N(C2=CC=CC=N2)CCN3CCN(C4=CC=CC=C4OC)CC3)=O)C=C1
Molekylvikt (g/mol) 434.513646