Learn More
p-Nitrobenzoyl-2,3,4-tri-O-benzyl-Alpha,Beta-L-fucopyranose, TRC
Product Code. 30468794
Change view
Click to view available options
Kvantitet:
500 mg
Product Code. Kvantitet
30468794 500 mg
1 options
Product Code. 30468794 Supplier Toronto Research Chemicals Supplier No. TRCN495000500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material p-Nitrobenzoyl-2,3,4-tri-O-benzyl-a,b-L-fucopyranose
CAS 151909-88-5
Molekylformel C34 H33 N O8
Kvantitet 500 mg
InChI formel InChI=1S/C34H33NO8/c1-24-30(39-21-25-11-5-2-6-12-25)31(40-22-26-13-7-3-8-14-26)32(41-23-27-15-9-4-10-16-27)34(42-24)43-33(36)28-17-19-29(20-18-28)35(37)38/h2-20,24,30-32,34H,21-23H2,1H3/t24-,30+,31+,32-,34?/m0/s1
LEDER C[C@@H]1OC(OC(=O)c2ccc(cc2)[N+](=O)[O-])[C@@H](OCc3ccccc3)[C@H](OCc4ccccc4)[C@@H]1OCc5ccccc5
IUPAC-namn [(3S,4R,5R,6S)-6-methyl-3,4,5-tris(phenylmethoxy)oxan-2-yl] 4-nitrobenzoate
Molekylvikt (g/mol) 583.63
Formel vikt 583.2206