Läs mer
p-Nitrophenyl-2,3,4,6-Tetra-O-acetyl-Beta-D-glucopyranoside, TRC
Produktkod. 30481005
Change view
Kvantitet:
1 g
2 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481005 5 g
30466897 1 g
30477117 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30481005 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN5057405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5987-78-0
Molekylformel C20H23NO12
Molekylvikt (g/mol) 469.4
Synonym p-Nitrophenyl beta-D-Glucopyranoside 2,3,4,6-Tetraacetate
IUPAC-namn [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-(4-nitrophenoxy)oxan-2-yl]methyl acetate
LEDER CC(=O)OC[C@H]1O[C@@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
Specifikationer
Kemiskt namn eller material p-Nitrophenyl-2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranoside
Kvantitet 5 g
InChI formel InChI=1S/C20H23NO12/c1-10(22)28-9-16-17(29-11(2)23)18(30-12(3)24)19(31-13(4)25)20(33-16)32-15-7-5-14(6-8-15)21(26)27/h5-8,16-20H,9H2,1-4H3/t16-,17-,18+,19-,20-/m1/s1
Formel vikt 469.122