Läs mer
p-Nitrophenyl 2-Acetamido-2-deoxy-3,4,6-tri-O-acetyl-Alpha-D-glucopyranoside, TRC
Produktkod. 30468789
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30468789 25 mg
30468788 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30468789 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN50209525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-Nitrophenyl 2-Acetamido-2-deoxy-3,4,6-tri-O-acetyl-a-D-glu
CAS 13089-23-1
Molekylformel C20 H24 N2 O11
Kvantitet 25 mg
InChI formel InChI=1S/C20H24N2O11/c1-10(23)21-17-19(31-13(4)26)18(30-12(3)25)16(9-29-11(2)24)33-20(17)32-15-7-5-14(6-8-15)22(27)28/h5-8,16-20H,9H2,1-4H3,(H,21,23)/t16-,17-,18-,19-,20+/m1/s1
LEDER CC(=O)N[C@H]1[C@@H](Oc2ccc(cc2)[N+](=O)[O-])O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn [(2R,3S,4R,5R,6R)-5-acetamido-3,4-diacetyloxy-6-(4-nitrophenoxy)oxan-2-yl]methyl acetate
Molekylvikt (g/mol) 468.41
Formel vikt 468.138