Läs mer
p-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(Beta-D-galactopyranosyl)-Beta-D-glucopyranoside, TRC
Produktkod. 30479092
Change view
Kvantitet:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479092 25 mg
30476139 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30479092 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN50125525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74211-28-2
Molekylformel C20 H28 N2 O13
Molekylvikt (g/mol) 504.44
IUPAC-namn N-[(2S,3R,4R,5S,6R)-4-hydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)-5-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-3-yl]acetamide
LEDER CC(=O)N[C@@H]1[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@@H](CO)O[C@H]1Oc3ccc(cc3)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material p-Nitrophenyl 2-Acetamido-2-deoxy-4-O-(beta-D-galactopyranosyl)-beta-D-glucopyranoside
Kvantitet 25 mg
InChI formel InChI=1S/C20H28N2O13/c1-8(25)21-13-15(27)18(35-20-17(29)16(28)14(26)11(6-23)33-20)12(7-24)34-19(13)32-10-4-2-9(3-5-10)22(30)31/h2-5,11-20,23-24,26-29H,6-7H2,1H3,(H,21,25)/t11-,12-,13-,14+,15-,16+,17-,18-,19-,20+/m1/s1
Formel vikt 504.1591