Läs mer
p-Nitrophenyl 2-Acetamido-2-deoxy-Alpha-D-galactopyranoside, TRC
Produktkod. 30480317
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480317 10 mg
30470687 25 mg
30475888 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480317 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN50000010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23646-68-6
Molekylformel C14 H18 N2 O8
Molekylvikt (g/mol) 342.3
IUPAC-namn N-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
LEDER CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@@H]1Oc2ccc(cc2)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material p-Nitrophenyl 2-Acetamido-2-deoxy-a-D-galactopyranoside
Kvantitet 10 mg
InChI formel InChI=1S/C14H18N2O8/c1-7(18)15-11-13(20)12(19)10(6-17)24-14(11)23-9-4-2-8(3-5-9)16(21)22/h2-5,10-14,17,19-20H,6H2,1H3,(H,15,18)/t10-,11-,12+,13-,14+/m1/s1
Formel vikt 342.1063