missing translation for 'onlineSavingsMsg'
Learn More

p-Nitrophenyl 2-Azido-2-deoxy-Alpha-D-galactopyranoside, TRC

Product Code. 30471215
Change view
Click to view available options
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30471215 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471215 Supplier Toronto Research Chemicals Supplier No. TRCN5030005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material p-Nitrophenyl 2-Azido-2-deoxy-a-D-galactopyranoside
CAS 210418-04-5
Molekylformel C12 H14 N4 O7
Kvantitet 5 mg
InChI formel InChI=1S/C12H14N4O7/c13-15-14-9-11(19)10(18)8(5-17)23-12(9)22-7-3-1-6(2-4-7)16(20)21/h1-4,8-12,17-19H,5H2/t8-,9-,10+,11-,12+/m1/s1
LEDER OC[C@H]1O[C@H](Oc2ccc(cc2)[N+](=O)[O-])[C@H](N=[N+]=[N-])[C@@H](O)[C@H]1O
IUPAC-namn (2R,3R,4R,5R,6R)-5-azido-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4-diol
Molekylvikt (g/mol) 326.26
Formel vikt 326.0862
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.