Läs mer
p-Nitrophenyl-a-D-maltoside, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | p-Nitrophenyl-a-D-maltoside |
| CAS | 17400-77-0 |
| Molekylformel | C18H25NO13 |
| Kvantitet | 200 mg |
| Synonym | α-D-Glucopyranoside','4-nitrophenyl 4-O-α-D-glucopyranosyl-, Glucopyranoside','p-nitrophenyl 4-O-α-D-glucopyranosyl-','α-D- (8CI), Maltoside','p-nitrophenyl (7CI), α-Maltoside','p-nitrophenyl (6CI), 4-Nitrophenyl 4-O-α-D-glucopyranosyl-α-D-glucopyranoside, 4-Nitrophenyl α-D-maltoside, p-Nitrophenyl α-D-maltoside |
| InChI formel | InChI=1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2/t9-,10-,11-,12+,13-,14-,15-,16-,17+,18-/m1/s1 |
| LEDER | OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](Oc3ccc(cc3)[N+](=O)[O-])O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC-namn | (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molekylvikt (g/mol) | 463.39 |
| Formel vikt | 463.1326 |