Läs mer
p-Nitrophenyl Acetate-13C,d3, TRC
Produktkod. 30180227
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180227 50 mg
30180199 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180227 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49804250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1794766-80-5
Molekylformel C713CH4D3NO4
Molekylvikt (g/mol) 185.16
Synonym p-Nitro-phenol Acetate-13C,d3, 4-Nitrophenyl Acetate-13C,d3, NSC 2633-13C,d3, p-Acetoxynitrobenzene-13C,d3, p-Nitrobenzene Acetate-13C,d3, p-Nitrophenol Acetate-13C,d3;
LEDER [2H]C([2H])([2H])[13C](=O)OC1=CC=C(C=C1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material p-Nitrophenyl Acetate-13C,d3
Kvantitet 50 mg
InChI formel InChI=1S/C8H7NO4/c1-6(10)13-8-4-2-7(3-5-8)9(11)12/h2-5H,1H3/i1D3,6+1
Formel vikt 185.0597