Läs mer
p-Nitrophenyl Beta-D-Cellobioside, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | p-Nitrophenyl beta-D-Cellobioside |
| CAS | 3482-57-3 |
| Molekylformel | C18 H25 N O13 |
| Kvantitet | 100 mg |
| Synonym | β-D-Glucopyranoside','4-nitrophenyl 4-O-β-D-glucopyranosyl-, Cellobioside','p-nitrophenyl (7CI), Cellobioside','p-nitrophenyl','β-D- (8CI), 4-Nitrophenyl 4-O-β-D-glucopyranosyl-β-D-glucopyranoside, p-Nitrophenyl cellobioside, p-Nitrophenyl β-D-cellobioside, p-Nitrophenyl β-cellobioside |
| InChI formel | InChI=1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2/t9-,10-,11-,12+,13-,14-,15-,16-,17-,18+/m1/s1 |
| LEDER | OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](Oc3ccc(cc3)[N+](=O)[O-])O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC-namn | (2S,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molekylvikt (g/mol) | 463.39 |
| Formel vikt | 463.1326 |