Läs mer
p-Nitrophenyl Beta-D-Cellopentaoside, TRC
Produktkod. 30476317
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476317 10 mg
30478027 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30476317 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN50310010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 129411-63-8
Molekylformel C36 H55 N O28
Molekylvikt (g/mol) 949.81
IUPAC-namn (2S,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-[(2R,4R,5R,6S)-6-[(2R,3S,4R,5R,6S)-6-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
LEDER OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](OC3[C@H](O)[C@@H](O)[C@H](O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@H]5[C@H](O)[C@@H](O)[C@H](Oc6ccc(cc6)[N+](=O)[O-])O[C@@H]5CO)O[C@@H]4CO)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material p-Nitrophenyl beta-D-Cellopentaoside
Kvantitet 10 mg
InChI formel InChI=1S/C36H55NO28/c38-5-12-17(43)18(44)23(49)33(57-12)62-29-14(7-40)59-35(25(51)20(29)46)64-31-16(9-42)61-36(27(53)22(31)48)65-30-15(8-41)60-34(26(52)21(30)47)63-28-13(6-39)58-32(24(50)19(28)45)56-11-3-1-10(2-4-11)37(54)55/h1-4,12-36,38-53H,5-9H2/t12-,13-,14-,15-,16-,17-,18+,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31?,32-,33+,34+,35+,36+/m1/s1
Formel vikt 949.2911