Läs mer
P,P'-[(1,2-Phenylene)bis(methylene)]bisphosphonic Acid P,P,P',P'-tetraethyl ester, TRC
Produktkod. 30416672
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30416672 10 g
1 options
Produktkod. 30416672 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP31981510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material P,P'-[(1,2-Phenylene)bis(methylene)]bisphosphonic Acid P,P,P',P'-tetraethyl ester
CAS 42092-05-7
Molekylformel C16H28O6P2
Kvantitet 10 g
Synonym (o-Phenylenedimethylene)diphosphonic Acid Tetraethyl Ester, [1,2-Phenylenebis(methylene)]bisphosphonic Acid Tetraethyl Ester, Tetraethyl o-Xylylenediphosphonate
InChI formel InChI=1S/C16H28O6P2/c1-5-19-23(17,20-6-2)13-15-11-9-10-12-16(15)14-24(18,21-7-3)22-8-4/h9-12H,5-8,13-14H2,1-4H3
LEDER CCOP(=O)(Cc1ccccc1CP(=O)(OCC)OCC)OCC
IUPAC-namn 1,2-bis(diethoxyphosphorylmethyl)benzene
Molekylvikt (g/mol) 378.34
Formel vikt 378.1361