Läs mer
p,p'-Dimethoxytriphenylcarbinol, TRC
Produktkod. 30372498
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30372498 500 mg
1 options
Produktkod. 30372498 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD470470500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p,p'-Dimethoxytriphenylcarbinol
CAS 40615-35-8
Molekylformel C21H20O3
Kvantitet 500 mg
Synonym alpha,alpha-Bis(p-methoxyphenyl)benzyl alcohol, 4,4'-Dimethoxytrityl alcoholBis(4-methoxyphenyl)phenylmethanol, DMT-OH, 4-Methoxy-alpha-(4-methoxyphenyl)-alpha-phenylbenzenemethanol
InChI formel InChI=1S/C21H20O3/c1-23-19-12-8-17(9-13-19)21(22,16-6-4-3-5-7-16)18-10-14-20(24-2)15-11-18/h3-15,22H,1-2H3
LEDER COc1ccc(cc1)C(O)(c2ccccc2)c3ccc(OC)cc3
Molekylvikt (g/mol) 320.39
Formel vikt 320.1412