Läs mer
p-Phenyl-p-(2,4,6-trimethylbenzoyl)phosphinic Acid Ethyl Ester, TRC
Produktkod. 17915105
Change view
Kvantitet:
1 g
5 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17915105 1 g
17921265 100 g
17944985 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17915105 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3376001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 84434-11-7
Molekylformel C18 H21 O3 P
Molekylvikt (g/mol) 316.33
Synonym Phosphinic acid, phenyl(2,4,6-trimethylbenzoyl)-, ethyl ester (9CI), 2,4,6-Trimethylbenzoylethoxyphenylphosphine oxide, 2,4,6-Trimethylbenzoylphenylethoxyphosphine oxide, Darocur TPO-L, Ethyl (2,4,6-trimethylbenzoyl)phenylphosphinate, Ethyl phenyl(2,4,6-trimethylbenzoyl)phosphinate, Irgacure TPO-L, JRcure TPO-L, LR 8893X, Lucirin LR 8893X, Lucirin TPO Liquid, Lucirin TPO-L, Omnirad TPO-L, Speedcure TPO-L, TPO-L
IUPAC-namn [ethoxy(phenyl)phosphoryl]-(2,4,6-trimethylphenyl)methanone
LEDER CCOP(=O)(C(=O)c1c(C)cc(C)cc1C)c2ccccc2
Specifikationer
Kemiskt namn eller material 2,4,6-Trimethylbenzoylethoxyphenylphosphine
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C18H21O3P/c1-5-21-22(20,16-9-7-6-8-10-16)18(19)17-14(3)11-13(2)12-15(17)4/h6-12H,5H2,1-4H3
Formel vikt 316.1228
Procent renhet >95%