Läs mer
p-Salicylic Acid 4-Glucuronide Potassium Salt, TRC
Produktkod. 30423283
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30423283 50 mg
1 options
Produktkod. 30423283 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS08814050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material p-Salicylic Acid 4-Glucuronide Potassium Salt
CAS 1376574-47-8
Molekylformel C13H12K2O9
Kvantitet 50 mg
Synonym 4-Carboxyphenyl β-D-Glucopyranosiduronic Acid Potassium Salt, p-Carboxyphenyl glucosiduronic Acid Potassium Salt
InChI formel InChI=1S/C13H14O9.2K/c14-7-8(15)10(12(19)20)22-13(9(7)16)21-6-3-1-5(2-4-6)11(17)18;;/h1-4,7-10,13-16H,(H,17,18)(H,19,20);;/q;2*+1/p-2/t7-,8-,9+,10?,13+;;/m0../s1
LEDER O[C@H]1[C@H](O)[C@@H](O)[C@H](OC2=CC=C(C(O[K])=O)C=C2)O[C@@H]1C(O[K])=O
IUPAC-namn potassium (2S,3S,4S,5R,6S)-6-(4-carboxylatophenoxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylate
Molekylvikt (g/mol) 390.43
Formel vikt 389.98