Läs mer
p-SCN-Bn-DTPA Trihydrochloride Salt (>90%), TRC
Produktkod. 30008273
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008273 100 mg
17967634 50 mg
17917454 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008273 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS200105100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2614335-77-0
Molekylformel C22H28N4O10S • 3(HCl)
Molekylvikt (g/mol) 649.92
Synonym N-[2-[Bis(carboxymethyl)amino]ethyl]-N-[(2S)-2-[bis(carboxymethyl)amino]-3-(4-isothiocyanatophenyl)propyl]glycine Trihydrochloride, (S)-1-(4-Isothiocyanatobenzyl)diethylenetriamine-N,N,N',N' ',N' '-pentaacetic Acid Trihydrochloride;
IUPAC-namn (S)-2,2'-((2-((2-(bis(carboxymethyl)amino)-3-(4-isothiocyanatophenyl)propyl)(carboxymethyl)amino)ethyl)azanediyl)diacetic acid hydrochloride
LEDER OC(CN([C@@H](CC1=CC=C(C=C1)N=C=S)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O)CC(O)=O)=O.Cl
Specifikationer
Kemiskt namn eller material p-SCN-Bn-DTPA Trihydrochloride Salt
Kvantitet 100 mg
InChI formel InChI=1S/C22H28N4O10S.ClH/c27-18(28)9-24(5-6-25(10-19(29)30)11-20(31)32)8-17(26(12-21(33)34)13-22(35)36)7-15-1-3-16(4-2-15)23-14-37;/h1-4,17H,5-13H2,(H,27,28)(H,29,30)(H,31,32)(H,33,34)(H,35,36);1H/t17-;/m0./s1
Formel vikt 648.09