Läs mer
[p-Terphenyl]-4,4' '-dicarboxylic Acid, TRC
Produktkod. 30414298
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414298 500 mg
30417088 100 mg
30419573 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30414298 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT116700500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13653-84-4
Molekylformel C20 H14 O4
Molekylvikt (g/mol) 318.32
Synonym [1,1':4',1' '-Terphenyl]-4,4' '-dicarboxylic Acid
IUPAC-namn 4-[4-(4-carboxyphenyl)phenyl]benzoic acid
LEDER OC(=O)c1ccc(cc1)c2ccc(cc2)c3ccc(cc3)C(=O)O
Specifikationer
Kemiskt namn eller material [p-Terphenyl]-4,4' '-dicarboxylic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C20H14O4/c21-19(22)17-9-5-15(6-10-17)13-1-2-14(4-3-13)16-7-11-18(12-8-16)20(23)24/h1-12H,(H,21,22)(H,23,24)
Formel vikt 318.0892