Läs mer
p-Toluidine Trifluoroacetamide, TRC
Produktkod. 30455974
Change view
Kvantitet:
500 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455974 500 mg
30452065 1 g
30453686 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455974 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT536280500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 350-96-9
Molekylformel C9H8F3NO
Molekylvikt (g/mol) 203.16
Synonym p-Methyltrifluoroacetanilide, 2,2,2-Trifluoro-p-acetotoluidide, 2,2,2-Trifluoro-N-(4-methylphenyl)acetamide, NSC 87411
IUPAC-namn 2,2,2-trifluoro-N-(4-methylphenyl)acetamide
LEDER Cc1ccc(NC(=O)C(F)(F)F)cc1
Specifikationer
Kemiskt namn eller material p-Toluidine Trifluoroacetamide
Kvantitet 500 mg
InChI formel InChI=1S/C9H8F3NO/c1-6-2-4-7(5-3-6)13-8(14)9(10,11)12/h2-5H,1H3,(H,13,14)
Formel vikt 203.0558