missing translation for 'onlineSavingsMsg'
Learn More

p19 INK4d Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10272-2-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826553

  • 3313.85 SEK / 150µL
Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

p19 / INK 4d is a specific inhibitor of cdk 4 / cdk 6 and is periodically expressed during cell cycle. It is maximally induced as cells enter S-phase.
Specifications

Specifications

p19 INK4d
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P55273, Q60773
CDKN2D, INK4D, P19, p19 INK4D, p19 INK4d
Rabbit
IgG
150 μL
-20°C
Primary
1032, 12581, 494444
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CDKN2D
Liquid
Cdkn2d
p19 INK4d Fusion Protein Ag0415
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers