missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2RY1 Mouse anti-Human, Unconjugated, Clone: 1A1F3, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | P2RY1 |
---|---|
Clone | 1A1F3 |
Concentration | 500 μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840406
|
Proteintech
67654-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830406
|
Proteintech
67654-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The P2RY1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human P2RY1. This antibody recognizes human, pig, mouse, rat, rabbit antigen. The P2RY1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
P2RY1 | |
500 μg/mL | |
Monoclonal | |
Mouse | |
5028 | |
Primary |
1A1F3 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Human, Mouse, Rat, Pig, Rabbit | |
IgG1 |