missing translation for 'onlineSavingsMsg'
Learn More

P3H1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26691-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16885085

  • 1211.96 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Basement membrane-associated chondroitin sulfate proteoglycan (CSPG). Has prolyl 3-hydroxylase activity catalyzing the post-translational formation of 3-hydroxyproline in -Xaa-Pro- Gly- sequences in collagens, especially types IV and V. May be involved in the secretory pathway of cells. Has growth suppressive activity in fibroblasts.
Specifications

Specifications

P3H1
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q32P28
GROS1, Growth suppressor 1, LEPRE1, Leprecan 1, P3H1, Prolyl 3 hydroxylase 1
Rabbit
IgG
20 μL
-20°C
Primary
64175
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
P3H1
Liquid
P3H1
P3H1 Fusion Protein Ag24904
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers