missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P4HA3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23185-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Catalyzes the post-translational formation of 4- hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins.Specifications
P4HA3 | |
Polyclonal | |
Unconjugated | |
P4HA3 | |
4 PH alpha 3, P4HA3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
283208, 320452 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6W3F0, Q7Z4N8 | |
P4HA3 | |
P4HA3 Fusion Protein Ag19716 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |