missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P62/SQSTM1 Mouse anti-Human, Clone: 1H5C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66184-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The molecular weight of p62 is predicted as 48/ 38 kDa, while western blot analyses using this antibody demonstrate the major band around 60-62 kDa in various tissues.
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.Specifications
P62/SQSTM1 | |
Monoclonal | |
1.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13501 | |
SQSTM1 | |
P62/SQSTM1 Fusion Protein Ag13131 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1H5C1 | |
Unconjugated | |
SQSTM1 | |
A170, EBIAP, ORCA, OSIL, p60, P62, P62;SQSTM1, p62B, PDB3, sequestosome 1, SQSTM1, Ubiquitin binding protein p62, ZIP3 | |
Mouse | |
Protein A | |
RUO | |
8878 | |
-20°C | |
Liquid |