Läs mer
PAC-1, TRC
Produktkod. 30414527
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414527 5 mg
30420214 10 mg
30415300 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30414527 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP1320005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 315183-21-2
Molekylformel C23H28N4O2
Molekylvikt (g/mol) 392.49
Synonym 4-(Phenylmethyl)-1-piperazineacetic Acid 2-[[2-Hydroxy-3-(2-propen-1-yl)phenyl] methylene]hydrazide, Procaspase Activating Compound 1
IUPAC-namn 2-(4-benzylpiperazin-1-yl)-N-[(E)-(2-hydroxy-3-prop-2-enylphenyl)methylideneamino]acetamide
LEDER Oc1c(CC=C)cccc1\C=N\NC(=O)CN2CCN(Cc3ccccc3)CC2
Specifikationer
Kemiskt namn eller material PAC-1
Kvantitet 5 mg
InChI formel InChI=1S/C23H28N4O2/c1-2-7-20-10-6-11-21(23(20)29)16-24-25-22(28)18-27-14-12-26(13-15-27)17-19-8-4-3-5-9-19/h2-6,8-11,16,29H,1,7,12-15,17-18H2,(H,25,28)/b24-16+
Formel vikt 392.2212