Läs mer
Paclitaxel Succinate, TRC
Produktkod. 30422414
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422414 250 mg
30414057 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422414 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP132530250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 117527-50-1
Molekylformel C51H55NO17
Molekylvikt (g/mol) 953.98
Synonym 2'-O-Succinoylpaclitaxel, 2'-O-Succinylpaclitaxel, 2'-Succinylpaclitaxel, 2'-Succinyltaxol, Paclitaxel 2'-(Hydrogen Succinate), Paclitaxel 2'-Hemisuccinate, [2aR-[2aα,4β,4aβ,6β,9α(1R*,2S*),11α,12α,12aα,12bα]]-Mono[2-(benzoylamino)-1-[[[6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl]oxy]carbonyl]-2-phenylethyl] Ester Butanedioic Acid, Mono[(1R,2S)-2-(benzoylamino)-1-[[[(2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl]oxy]carbonyl]-2-phenylethyl] Ester Butanedioic Acid
IUPAC-namn 4-(((1S,2R)-1-benzamido-3-(((2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-diacetoxy-12-(benzoyloxy)-4,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-1H-7,11-methanocyclodeca[3,4]benzo[1,2-b]oxet-9-yl)oxy)-3-oxo-1-phenylpropan-2-yl)oxy)-4-oxobutanoic acid
LEDER O=C(C)O[C@H](C1=C(C)[C@@H](OC([C@H](OC(CCC(O)=O)=O)[C@@H](NC(C2=CC=CC=C2)=O)C3=CC=CC=C3)=O)C[C@]([C@H](OC(C4=CC=CC=C4)=O)[C@@]5([H])[C@@]6(C)[C@@H](O)CC7C5(OC(C)=O)CO7)(O)C1(C)C)C6=O
Specifikationer
Kemiskt namn eller material Paclitaxel Succinate
Kvantitet 250 mg
InChI formel InChI=1S/C51H55NO17/c1-27-33(66-47(62)41(67-37(58)23-22-36(56)57)39(30-16-10-7-11-17-30)52-45(60)31-18-12-8-13-19-31)25-51(63)44(68-46(61)32-20-14-9-15-21-32)42-49(6,34(55)24-35-50(42,26-64-35)69-29(3)54)43(59)40(65-28(2)53)38(27)48(51,4)5/h7-21,33-35,39-42,44,55,63H,22-26H2,1-6H3,(H,52,60)(H,56,57)/t33-,34-,35+,39-,40+,41+,42-,44-,49+,50-,51+/m0/s1
Formel vikt 953.35