Läs mer
Paeoniflorin (>80%), TRC
Produktkod. 30427220
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427220 250 mg
30425031 100 mg
30421800 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427220 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP133830250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23180-57-6
Molekylformel C23 H28 O11
Molekylvikt (g/mol) 480.46
Synonym β-D-Glucopyranoside, (1aR,2S,3aR,5R,5aR,5bS)-5b-[(benzoyloxy)methyl]tetrahydro-5-hydroxy-2-methyl-2,5-methano-1H-3,4-dioxacyclobuta[cd]pentalen-1a(2H)-yl, Peoniflorin (7CI,8CI), β-D-Glucopyranoside, 5b-[(benzoyloxy)methyl]tetrahydro-5-hydroxy-2-methyl-2,5-methano-1H-3,4-dioxacyclobuta[cd]pentalen-1a(2H)-yl, [1aR-(1aα,2β,3aα,5α,5aα,5bα)]-, 2,5-Methano-1H-3,4-dioxacyclobuta[cd]pentalene, β-D-glucopyranoside deriv., (1aR,2S,3aR,5R,5aR,5bS)-5b-[(Benzoyloxy)methyl]tetrahydro-5-hydroxy-2-methyl-2,5-methano-1H-3,4-dioxacyclobuta[cd]pentalen-1a(2H)-yl β-D-glucopyranoside, NSC 178886, Paeoniflorin
LEDER C[C@@]12C[C@@]3(O)O[C@@H](O1)[C@]4(COC(=O)c5ccccc5)[C@H]3C[C@]24O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O
Specifikationer
Kemiskt namn eller material Paeoniflorin
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C23H28O11/c1-20-9-22(29)13-7-23(20,32-18-16(27)15(26)14(25)12(8-24)31-18)21(13,19(33-20)34-22)10-30-17(28)11-5-3-2-4-6-11/h2-6,12-16,18-19,24-27,29H,7-10H2,1H3/t12-,13-,14-,15+,16-,18+,19-,20+,21+,22-,23+/m1/s1
Formel vikt 480.1632
Procent renhet >95%