missing translation for 'onlineSavingsMsg'
Learn More

PAFAH1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14729-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823524

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Platelet-activating factor (PAF) is a biologically active phospholipid with diverse biologic effects. PAF is degraded to inactive products by hydrolysis of the acetyl group at the sn-2 position to produce the biologically inactive products LYSO-PAF and acetate. This reaction is catalyzed by PAF acetylhydrolase (PAFAH). The various monomeric and multimeric forms of the enzyme are composed of alpha, beta, and gamma PAFAH subunits.
Specifications

Specifications

PAFAH1B2
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35264, P68402, Q61206
PAF AH 30 kDa subunit, PAF AH subunit beta, PAFAH subunit beta, PAFAH1B2, PAFAHB
Rabbit
IgG
20 μL
-20°C
Primary
18475, 5049, 64189
Immunoprecipitation, Western Blot
Unconjugated
PAFAH1B2
Liquid
Pafah1b2
PAFAH1B2 Fusion Protein Ag6489
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers