missing translation for 'onlineSavingsMsg'
Learn More

PAK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21401-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881005

  • 4721.67 SEK / 150µL
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology. Alternativelt spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

PAK1
Polyclonal
Unconjugated
PAK1
Alpha PAK, p21 activated kinase 1, p65 PAK, PAK 1, PAK1, PAKalpha
Rabbit
Antigen Affinity Chromatography
RUO
18479, 29431, 5058
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.67 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88643, P35465, Q13153
Pak1
PAK1 Fusion Protein Ag16102
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers