missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21401-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
PAK proteins are critical effectors that link RhoGTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, include PAK1, PAK2, PAK3 and PAK4. These proteins serve as targets for the small GTP binding proteins Cdc42 and Rac and have been implicated in a wide range of biological activities. PAK1 regulates cell motility and morphology. Alternativelt spliced transcript variants encoding different isoforms have been found for this gene.Specifications
PAK1 | |
Polyclonal | |
Unconjugated | |
PAK1 | |
Alpha PAK, p21 activated kinase 1, p65 PAK, PAK 1, PAK1, PAKalpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18479, 29431, 5058 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88643, P35465, Q13153 | |
Pak1 | |
PAK1 Fusion Protein Ag16102 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |