missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27942-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the p21-activated kinase (PAK) family. The proteins of this family are Rac/Cdc42-associated Ste20-like Ser/Thr protein kinases, characterized by a highly conserved amino-terminal Cdc42/Rac interactive binding (CRIB) domain and a carboxyl-terminal kinase domain. PAK kinases are implicated in the regulation of a number of cellular processes, including cytoskeleton rearrangement, apoptosis and the MAP kinase signaling pathway. The protein encoded by this gene was found to interact with androgen receptor (AR), which is a steroid hormone-dependent transcription factor that is important for male sexual differentiation and development. This gene was found to be highly expressed in testis and prostate tissues and the encoded protein was shown to cotranslocate into the nucleus with AR in response to androgen. Alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
PAK6 | |
Polyclonal | |
Unconjugated | |
PAK6 | |
p21 activated kinase 6, PAK 5, PAK 6, PAK5, PAK6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214230, 296078, 56924 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3ULB5, Q9NQU5 | |
PAK6 | |
PAK6 Fusion Protein Ag27687 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |