missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAK6 Recombinant Human Protein
PAK6 is a serine/threonine kinase.
18240.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_064553 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 4mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 79.9kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468443
|
Invitrogen™
PR5307A |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10482503
|
Invitrogen™
PV4032 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PAK6 is expressed in the testis and prostate. It associates with both the estrogen and androgen receptors.
Specifications
NP_064553 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 4mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
79.9kDa | |
RUO - research use only | |
PAK6, PAK5 | |
Human | |
His Tag | |
Frozen |