missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 10% on carbon paste, Type 755

Brand:  Alfa Aesar™ H37277.06

Additional Details : CAS Number : 7440-05-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15467747

  • 182.20 SEK / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
7440-05-3
10% on carbon paste, Type 755
MFCD03457879

Specifications

Palladium
7440-05-3
MFCD03457879
Dark gray
10% on carbon paste, Type 755
Solid
Odorless
5g
Videos
SDS