missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on carbon paste, A405028-5

Brand:  Alfa Aesar™ H36996.22

Additional Details : CAS Number : 7440-05-3 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15457727

  • 5765.61 SEK / 100g


Estimated Shipment date
from Supplier 03-08-2018
Add to basket

Specifications

Chemical Identifiers

Palladium
7440-05-3
5% on carbon paste, A405028-5
MFCD03457879

Specifications

Palladium
7440-05-3
Pd
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Odorless
100g
5% on carbon paste, A405028-5
Solid
MFCD03457879
[Pd]
106.42
CHEBI:33363
Dark gray
Videos
SDS