missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 5% on carbon paste, A405028-5

Brand:  Alfa Aesar™ H36996.06

Additional Details : CAS Number : 7440-05-3 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15437727

  • 531.81 SEK / 5g


Estimated Shipment
12-10-2020
Add to basket

Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
palladium
Pd
MFCD03457879
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363
[Pd]

Specifications

Palladium
7440-05-3
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Solid
Gray
5% on carbon paste, A405028-5
Pd
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
[Pd]
106.42
CHEBI:33363
Odorless
5g
Videos
SDS Documents