Learn More
Palladium, 8%, Platinum, 2%; on activated carbon powder, nominally 50% water wet, Thermo Scientific™
Catalog Number p-7051904
Click to view available options
Quantity:
2 g
10 g
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
on carbon, black, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
on carbon, black, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Selective hydrogenation of nitrates to hydroxylamines, reduction of nitrogen oxides to N2 with H2 in the presence of CO and CO2
TRUSTED_SUSTAINABILITY
Quantity 2 g
Chemical Name or Material Palladium, Platinum, Pd, 8%, Pt, 2% on activated carbon powdernominally 50% water wet