missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 8%, Platinum, 2%; on activated carbon powder, nominally 50% water wet

Brand:  Alfa Aesar™ 042208.04

Additional Details : Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 15433875

  • 459.09 SEK / 2g


In stock, ready to ship: 1
Add to basket

Description

Description

  • Selective hydrogenation of nitrates to hydroxylamines, reduction of nitrogen oxides to N2 with H2 in the presence of CO and CO2
Specifications

Chemical Identifiers

7440-05-3
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
Pd
MFCD01074898
on carbon, black, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
CHEBI:33363

Specifications

Palladium, Platinum
Pd, 8%, Pt, 2% on activated carbon powdernominally 50% water wet
Pd
on carbon, black, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
106.42
CHEBI:33363
2g
7440-05-3
MFCD01074898
KDLHZDBZIXYQEI-UHFFFAOYSA-N
23938
Videos
SDS Documents