missing translation for 'onlineSavingsMsg'
Learn More

PALLD/palladin Mouse anti-Human, Porcine, Rat, Clone: 2A1F6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66601-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16888625

  • 1656.73 SEK / 20µL
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin′s different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.
Specifications

Specifications

PALLD/palladin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P0C5E3, Q8WX93
Palld
PALLD,palladin Fusion Protein Ag1288
20 μL
Primary
Porcine, Rat, Human
Antibody
IgG2a
Western Blot, Immunohistochemistry (Paraffin)
2A1F6
Unconjugated
PALLD
CGI 151, KIAA0992, palladin, PALLD, PNCA1, Sarcoma antigen NY SAR 77, SIH002
Mouse
Protein A
RUO
100627292, 23022, 364558
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers