Läs mer
Palmitic Acid Methylamide, TRC
Produktkod. 30410742
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410742 1 g
30422393 250 mg
30426759 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30410742 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP1445081G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7388-58-1
Molekylformel C17H35NO
Molekylvikt (g/mol) 269.47
Synonym N-Methyl-hexadecanamide, N-Methylhexadecanamide, N-Methylpalmitamide
IUPAC-namn N-methylhexadecanamide
LEDER CCCCCCCCCCCCCCCC(=O)NC
Specifikationer
Kemiskt namn eller material Palmitic Acid Methylamide
Kvantitet 1 g
InChI formel InChI=1S/C17H35NO/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(19)18-2/h3-16H2,1-2H3,(H,18,19)
Formel vikt 269.2719