Läs mer
Palovarotene, TRC
Produktkod. 30422665
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422665 100 mg
30426669 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422665 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP165900100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 410528-02-8
Molekylformel C27 H30 N2 O2
Molekylvikt (g/mol) 414.54
Synonym Benzoic acid, 4-[(1E)-2-[5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-3-(1H-pyrazol-1-ylmethyl)-2-naphthalenyl]ethenyl]-, 4-[(1E)-2-[5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-3-(1H-pyrazol-1-ylmethyl)-2-naphthalenyl]ethenyl]benzoic acid, Palovarotene, R 667, Ro 3300074
IUPAC-namn 4-[(E)-2-[5,5,8,8-tetramethyl-3-(pyrazol-1-ylmethyl)-6,7-dihydronaphthalen-2-yl]ethenyl]benzoic acid
LEDER CC1(C)CCC(C)(C)c2cc(\C=C\c3ccc(cc3)C(=O)O)c(Cn4cccn4)cc12
Specifikationer
Kemiskt namn eller material Palovarotene
Kvantitet 100 mg
InChI formel InChI=1S/C27H30N2O2/c1-26(2)12-13-27(3,4)24-17-22(18-29-15-5-14-28-29)21(16-23(24)26)11-8-19-6-9-20(10-7-19)25(30)31/h5-11,14-17H,12-13,18H2,1-4H3,(H,30,31)/b11-8+
Formel vikt 414.2307