Läs mer
Pancuronium Bromide, TRC
Produktkod. 30137662
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137662 50 mg
30136971 100 mg
30008830 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137662 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP17850050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15500-66-0
Molekylformel C35 H60 N2 O4 . 2 Br
Molekylvikt (g/mol) 732.67
Synonym Pancuronium bromide, 1,1′-[3α,17β-Bis(acetyloxy)-5α-androstane-2β,16β-diyl]bis(1-methylpiperidinium) dibromide, Piperidinium, 1,1'-(3α,17β-dihydroxy-5α-androstan-2β,16β-ylene)bis[1-methyl-, dibromide, diacetate (8CI), 5α-Androstane-3α,17β-diol, 2β,16β-bis(1-methylpiperidinio)-, dibromide, diacetate (8CI), Androstane, piperidinium deriv., 1,1'-(3α,17β-Dihydroxy-5α-androstan-2β,16β-ylene)bis[1-methylpiperidinium] dibromide diacetate, 2β,16β-Dipiperidino-5α-androstane-3α,17β-diol diacetate dimethobromide, Bromurex, Mioblock, NA 97, NSC 293162, Org NA 97, Org-NE 35, Pancuronium, Pancuronium bromide, Pancuronium dibromide, Pavulon
IUPAC-namn [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-acetyloxy-10,13-dimethyl-2,16-bis(1-methylpiperidin-1-ium-1-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate;dibromide
LEDER [Br-].[Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(=O)C)[N+]5(C)CCCCC5)[C@@]2(C)C[C@@H]1[N+]6(C)CCCCC6
Specifikationer
Kemiskt namn eller material Pancuronium Bromide
Kvantitet 50 mg
InChI formel InChI=1S/C35H60N2O4.2BrH/c1-24(38)40-32-21-26-13-14-27-28(35(26,4)23-31(32)37(6)19-11-8-12-20-37)15-16-34(3)29(27)22-30(33(34)41-25(2)39)36(5)17-9-7-10-18-36;;/h26-33H,7-23H2,1-6H3;2*1H/q+2;;/p-2/t26-,27+,28-,29-,30-,31-,32-,33-,34-,35-;;/m0../s1
Formel vikt 730.292