Läs mer
Panobinostat(LBH-589), TRC
Produktkod. 30414768
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414768 250 mg
30411139 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30414768 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP180500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Panobinostat
CAS 404950-80-7
Molekylformel C21 H23 N3 O2
Kvantitet 250 mg
Synonym 2-Propenamide','N-hydroxy-3-[4-[[[2-(2-methyl-1H-indol-3-yl)ethyl]amino]methyl]phenyl]-','(2E)-, (2E)-N-Hydroxy-3-[4-[[[2-(2-methyl-1H-indol-3-yl)ethyl]amino]methyl]phenyl]-2-propenamide, Farydak, LBH 589, Panobinostat
InChI formel InChI=1S/C21H23N3O2/c1-15-18(19-4-2-3-5-20(19)23-15)12-13-22-14-17-8-6-16(7-9-17)10-11-21(25)24-26/h2-11,22-23,26H,12-14H2,1H3,(H,24,25)/b11-10+
LEDER Cc1[nH]c2ccccc2c1CCNCc3ccc(\C=C\C(=O)NO)cc3
IUPAC-namn (E)-N-hydroxy-3-[4-[[2-(2-methyl-1H-indol-3-yl)ethylamino]methyl]phenyl]prop-2-enamide
Molekylvikt (g/mol) 349.43
Formel vikt 349.179