Läs mer
Pantoprazole Dimer (>90%), TRC
Produktkod. 30137684
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137684 10 mg
30007381 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137684 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP18290010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2115779-15-0
Molekylformel C32H28F4N6O8S2
Molekylvikt (g/mol) 764.72
Synonym 6,6'-Bis(difluoromethoxy)-2,2'-bis(((3,4-dimethoxypyridin-2-yl)methyl)sulfinyl)-3H,3'H-5,5'-bibenzo[d]imidazolel, USP Pantoprazole Related Compound E;
IUPAC-namn 6-(difluoromethoxy)-5-[6-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]-1H-benzimidazol-5-yl]-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]-1H-benzimidazole
LEDER COc1ccnc(CS(=O)c2nc3cc(c(OC(F)F)cc3[nH]2)c4cc5nc([nH]c5cc4OC(F)F)S(=O)Cc6nccc(OC)c6OC)c1OC
Specifikationer
Kemiskt namn eller material Pantoprazole Dimer
Kvantitet 10 mg
InChI formel InChI=1S/C32H28F4N6O8S2/c1-45-23-5-7-37-21(27(23)47-3)13-51(43)31-39-17-9-15(25(49-29(33)34)11-19(17)41-31)16-10-18-20(12-26(16)50-30(35)36)42-32(40-18)52(44)14-22-28(48-4)24(46-2)6-8-38-22/h5-12,29-30H,13-14H2,1-4H3,(H,39,41)(H,40,42)
Formel vikt 764.1346