missing translation for 'onlineSavingsMsg'
Learn More

Pantoprazole sodium salt hydrate, Thermo Scientific Chemicals

Catalog Number p-7077859
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
718635-09-7
C16H14F2N3NaO4S
405.35
MFCD08704580
YNWDKZIIWCEDEE-UHFFFAOYNA-N
pantoprazole sodium, pantoprazole sodium hydrate, pantozol hydrate, protonix hydrate, c16h14f2n3nao4s.h2o, sodium pantoprazole 1-hydrate, pantoprazole sodium hydrate hplc, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methanesulfinyl-1-sodio-1,3-benzodiazole hydrate, 5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl sulfinyl-1h-benzimidazole sodium salt hydrate
23684923
sodium;5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]benzimidazol-1-ide;hydrate
[Na+].COC1=C(OC)C(CS(=O)C2=NC3=CC(OC(F)F)=CC=C3[N-]2)=NC=C1
This item is not returnable. View return policy
718635-09-7
C16H14F2N3NaO4S
405.35
MFCD08704580
YNWDKZIIWCEDEE-UHFFFAOYNA-N
pantoprazole sodium, pantoprazole sodium hydrate, pantozol hydrate, protonix hydrate, c16h14f2n3nao4s.h2o, sodium pantoprazole 1-hydrate, pantoprazole sodium hydrate hplc, 5-difluoromethoxy-2-3,4-dimethoxypyridin-2-yl methanesulfinyl-1-sodio-1,3-benzodiazole hydrate, 5-difluoromethoxy-2-3,4-dimethoxy-2-pyridinyl methyl sulfinyl-1h-benzimidazole sodium salt hydrate
23684923
sodium;5-(difluoromethoxy)-2-[(3,4-dimethoxypyridin-2-yl)methylsulfinyl]benzimidazol-1-ide;hydrate
[Na+].COC1=C(OC)C(CS(=O)C2=NC3=CC(OC(F)F)=CC=C3[N-]2)=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 151.0°C to 155.0°C
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information (1% in water) Clear solution
Formula Weight 405.35
Chemical Name or Material Pantoprazole sodium salt hydrate
compliance-icons
Product Identifier
  • Pantoprazole sodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'