Läs mer
Pantoprazole Sulfide N-Oxide, TRC
Produktkod. 30424476
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424476 25 mg
30416054 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30424476 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP18303025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 953787-51-4
Molekylformel C16 H15 F2 N3 O4 S
Molekylvikt (g/mol) 383.37
Synonym Pantoprazole Sulphide N-Oxide
IUPAC-namn 6-(difluoromethoxy)-2-[(3,4-dimethoxy-1-oxidopyridin-1-ium-2-yl)methylsulfanyl]-1H-benzimidazole
LEDER COc1cc[n+]([O-])c(CSc2nc3ccc(OC(F)F)cc3[nH]2)c1OC
Specifikationer
Kemiskt namn eller material Pantoprazole Sulfide N-Oxide
Kvantitet 25 mg
InChI formel InChI=1S/C16H15F2N3O4S/c1-23-13-5-6-21(22)12(14(13)24-2)8-26-16-19-10-4-3-9(25-15(17)18)7-11(10)20-16/h3-7,15H,8H2,1-2H3,(H,19,20)
Formel vikt 383.0751