Learn More
PAPOLB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12821-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16890734
  • 1355.00 SEK / 20µL
Estimated Shipment: 21-05-2024
to see stock.
Description
Description
Testis-specific poly(A) polymerase.
Specifications
Specifications
PAPOLB
Polyclonal
Unconjugated
PAPOLB
PAP beta, PAPOLB, PAPT, Poly(A) polymerase beta, TPAP
Rabbit
Antigen Affinity Chromatography
RUO
304300, 56522, 56903
-20°C
Liquid
Western Blot
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NRJ5, Q9WVP6
Papolb
PAPOLB Fusion Protein Ag3487
20 μL
Primary
Human, Rat, Mouse