Läs mer
Paprotrain, MedChemExpress
Produktkod. 16493328
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16493328 10 mg
16464576 100 mg
16459866 50 mg
16432726 25 mg
16426057 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16493328 Leverantör Medchem Express Leverantörsnummer HY101298
MedChemExpress Paprotrain is a cell-permeable inhibitor of the kinesin MKLP-2, inhibits the ATPase activity of MKLP-2 with an IC50 of 1.35 μM and a Ki of 3.36 μM and shows a moderate inhibition activity on DYRK1A with an IC50 of 5.5 μM.
Paprotrain is a cell-permeable inhibitor of the kinesin MKLP-2, inhibits the ATPase activity of MKLP-2 with an IC50 of 1.35 μM and a Ki of 3.36 μM and shows a moderate inhibition activity on DYRK1A with an IC50 of 5.5 μM.
Specifikationer
Kemiskt namn eller material Paprotrain
Fysisk form Solid
CAS 57046-73-8
Färg Yellow
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C16H11N3
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation DMSO : ≥ 100 mg/mL (407.70 mM)
LEDER N#C/C(C1=CNC2=CC=CC=C21)=C\C3=CN=CC=C3
Molekylvikt (g/mol) 245.28
Formel vikt 245.28
Procent renhet 99.54%